Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTGCAGCCATTCCTCTTTGTTG[A/G]TGTAGGGGAGGAGAGAAGAGGTCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180231 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRABP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRABP2 - cellular retinoic acid binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199723.1 | 825 | UTR 3 | NP_001186652.1 | |||
NM_001878.3 | 825 | UTR 3 | NP_001869.1 | |||
XM_017000343.1 | 825 | UTR 3 | XP_016855832.1 |