Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGATCCAGGAGATCCAATAGC[A/G]TAGTTTCTTTCAGTCCATATGCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605877 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FCRL5 - Fc receptor like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195388.1 | 5274 | UTR 3 | NP_001182317.1 | |||
NM_031281.2 | 5274 | UTR 3 | NP_112571.2 | |||
XM_011510030.1 | 5274 | Intron | XP_011508332.1 | |||
XM_011510031.1 | 5274 | Intron | XP_011508333.1 | |||
XM_011510032.1 | 5274 | Intron | XP_011508334.1 | |||
XM_011510033.1 | 5274 | Intron | XP_011508335.1 |
Set Membership: |
HapMap |