Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACATTTCACATTGTATTAAACC[C/T]GGCATATCTTATCTCTCCTGCTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164873 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYCSP52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYCSP52 - cytochrome c, somatic pseudogene 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETV3 - ETS variant 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145312.1 | Intron | NP_001138784.1 | ||||
NM_005240.1 | Intron | NP_005231.1 | ||||
XM_006711210.2 | Intron | XP_006711273.1 |
Set Membership: |
HapMap |