Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGACCGAGAGCGCCGGCCGCACGG[A/C]GGACAAGTCGCCGCGCCAGCAGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SYPL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYPL2 - synaptophysin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040709.1 | 398 | Missense Mutation | GAG,GCG | E,A 11 | NP_001035799.1 | |
XM_011541283.2 | 398 | Missense Mutation | GAG,GCG | E,A 11 | XP_011539585.1 | |
XM_011541284.2 | 398 | Missense Mutation | GAG,GCG | E,A 11 | XP_011539586.1 | |
XM_011541285.1 | 398 | Missense Mutation | GAG,GCG | E,A 11 | XP_011539587.1 |