Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTAAAGGATATGAGAGATTATT[A/C]CAAAGATACCTACTTTACAGTATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611298 | ||||||||||||||||||||
Literature Links: |
C1orf194 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf194 - chromosome 1 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1324 - KIAA1324 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267048.1 | Intron | NP_001253977.1 | ||||
NM_001284352.1 | Intron | NP_001271281.1 | ||||
NM_001284353.1 | Intron | NP_001271282.1 | ||||
NM_020775.4 | Intron | NP_065826.2 | ||||
XM_011541825.1 | Intron | XP_011540127.1 | ||||
XM_011541826.2 | Intron | XP_011540128.1 | ||||
XM_011541827.1 | Intron | XP_011540129.1 | ||||
XM_017001883.1 | Intron | XP_016857372.1 |