Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612818 MIM: 606020 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUSAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUSAP1 - nucleolar and spindle associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243142.1 | 215 | Missense Mutation | ACC,GCC | T,A 33 | NP_001230071.1 | |
NM_001243143.1 | 215 | Missense Mutation | ACC,GCC | T,A 33 | NP_001230072.1 | |
NM_001243144.1 | 215 | Intron | NP_001230073.1 | |||
NM_001301136.1 | 215 | Missense Mutation | ACC,GCC | T,A 33 | NP_001288065.1 | |
NM_016359.4 | 215 | Missense Mutation | ACC,GCC | T,A 33 | NP_057443.2 | |
NM_018454.7 | 215 | Missense Mutation | ACC,GCC | T,A 33 | NP_060924.4 | |
XM_005254428.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_005254485.1 | |
XM_005254430.4 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_005254487.1 | |
XM_005254431.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_005254488.1 | |
XM_006720559.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_006720622.1 | |
XM_006720560.3 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_006720623.1 | |
XM_006720561.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_006720624.1 | |
XM_006720562.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_006720625.1 | |
XM_006720563.2 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_006720626.1 | |
XM_017022294.1 | 215 | Missense Mutation | ACC,GCC | T,A 33 | XP_016877783.1 | |
XM_017022295.1 | 215 | UTR 5 | XP_016877784.1 |
OIP5 - Opa interacting protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |