Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTATTTTGGTCTTCGGCCACCAC[A/G]GGGCATTTCCTTCCAACCTAATTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616786 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPKBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAPKBP1 - mitogen-activated protein kinase binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128608.1 | Intron | NP_001122080.1 | ||||
NM_001265611.1 | Intron | NP_001252540.1 | ||||
NM_014994.2 | Intron | NP_055809.2 | ||||
XM_006720438.1 | Intron | XP_006720501.1 | ||||
XM_006720439.2 | Intron | XP_006720502.1 | ||||
XM_011521383.1 | Intron | XP_011519685.1 | ||||
XM_011521384.2 | Intron | XP_011519686.1 | ||||
XM_017022017.1 | Intron | XP_016877506.1 |