Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGAACAAGAACATAGCTACATG[C/T]TTAAAGTTATTGTTTTGAAAGGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608700 | ||||||||||||||||||||
Literature Links: |
MIR5697 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR5697 - microRNA 5697 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMNAT1 - nicotinamide nucleotide adenylyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297778.1 | Intron | NP_001284707.1 | ||||
NM_001297779.1 | Intron | NP_001284708.1 | ||||
NM_022787.3 | Intron | NP_073624.2 | ||||
XM_011541971.2 | Intron | XP_011540273.1 | ||||
XM_017002107.1 | Intron | XP_016857596.1 | ||||
XM_017002108.1 | Intron | XP_016857597.1 |