Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCCTGTCCCTGCCTCCCCAC[A/C]CCCCCCATCCCCAAGTACTTACGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300061 | ||||||||||||||||||||
Literature Links: |
ZMYM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZMYM3 - zinc finger MYM-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171162.1 | Intron | NP_001164633.1 | ||||
NM_001171163.1 | Intron | NP_001164634.1 | ||||
NM_005096.3 | Intron | NP_005087.1 | ||||
NM_201599.2 | Intron | NP_963893.1 | ||||
XM_005262309.3 | Intron | XP_005262366.1 | ||||
XM_005262310.2 | Intron | XP_005262367.1 | ||||
XM_011531062.2 | Intron | XP_011529364.1 |