Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAGGCCTTAGGGACAAGCGGA[C/T]GTGGTGAGGGTGTCAGGGTGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608544 MIM: 602898 MIM: 602399 MIM: 610053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC10 - histone deacetylase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK11 - mitogen-activated protein kinase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK12 - mitogen-activated protein kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303252.1 | Intron | NP_001290181.1 | ||||
NM_002969.4 | Intron | NP_002960.2 |
TUBGCP6 - tubulin gamma complex associated protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |