Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAAAATAAACTCAGGGGCTAT[A/G]TTTCATTCAACCGAACAAGACAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHBDL3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RHBDL3 - rhomboid like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138328.2 | Intron | NP_612201.1 | ||||
XM_006721733.3 | Intron | XP_006721796.1 | ||||
XM_006721734.3 | Intron | XP_006721797.1 | ||||
XM_011524443.2 | Intron | XP_011522745.1 | ||||
XM_017024272.1 | Intron | XP_016879761.1 | ||||
XM_017024273.1 | Intron | XP_016879762.1 | ||||
XM_017024274.1 | Intron | XP_016879763.1 | ||||
XM_017024275.1 | Intron | XP_016879764.1 | ||||
XM_017024276.1 | Intron | XP_016879765.1 | ||||
XM_017024277.1 | Intron | XP_016879766.1 | ||||
XM_017024278.1 | Intron | XP_016879767.1 | ||||
XM_017024279.1 | Intron | XP_016879768.1 | ||||
XM_017024280.1 | Intron | XP_016879769.1 |