Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACATCATTACTAGGGGGCTTTTG[A/G]CCATGATATGCTCAGGGTCCTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 301300 | ||||||||||||||||||||
Literature Links: |
ALAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALAS2 - 5'-aminolevulinate synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000032.4 | Intron | NP_000023.2 | ||||
NM_001037967.3 | Intron | NP_001033056.1 | ||||
NM_001037968.3 | Intron | NP_001033057.1 | ||||
XM_005261995.3 | Intron | XP_005262052.1 | ||||
XM_011530771.1 | Intron | XP_011529073.1 | ||||
XM_017029354.1 | Intron | XP_016884843.1 |
PAGE2B - PAGE family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015038.2 | Intron | NP_001015038.1 | ||||
XM_011530785.2 | Intron | XP_011529087.1 | ||||
XM_011530786.2 | Intron | XP_011529088.1 | ||||
XM_011530787.2 | Intron | XP_011529089.1 | ||||
XM_017029513.1 | Intron | XP_016885002.1 |