Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACCTCTTACATCGATCTCTTG[A/G]TAGGAGGGGTTTTCTGTGTTTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615133 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNTL5 - polypeptide N-acetylgalactosaminyltransferase-like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145292.3 | Intron | NP_660335.2 | ||||
XM_006715862.3 | Intron | XP_006715925.1 | ||||
XM_011515849.2 | Intron | XP_011514151.1 | ||||
XM_011515852.2 | Intron | XP_011514154.1 | ||||
XM_011515853.2 | Intron | XP_011514155.1 | ||||
XM_017011793.1 | Intron | XP_016867282.1 | ||||
XM_017011794.1 | Intron | XP_016867283.1 | ||||
XM_017011795.1 | Intron | XP_016867284.1 | ||||
XM_017011796.1 | Intron | XP_016867285.1 |
Set Membership: |
HapMap |