Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGTGGAGATGGTGTTTCACTG[G/T]GTTGTCCCAGGCTGGTCTGAAACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180740 | ||||||||||||||||||||
Literature Links: |
SNRNP70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNRNP70 - small nuclear ribonucleoprotein U1 subunit 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301069.1 | Intron | NP_001287998.1 | ||||
NM_003089.5 | Intron | NP_003080.2 | ||||
XM_005259178.1 | Intron | XP_005259235.1 | ||||
XM_011527240.2 | Intron | XP_011525542.2 | ||||
XM_011527241.2 | Intron | XP_011525543.1 |