Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCATTTTAATAATGTAATAATC[C/T]CTCTATAAGTCTCAGTTTCCAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121013 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GJA5 - gap junction protein alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005266.6 | 4681 | UTR 3 | NP_005257.2 | |||
NM_181703.3 | 4681 | UTR 3 | NP_859054.1 | |||
XM_005272951.4 | 4681 | UTR 3 | XP_005273008.1 | |||
XM_017001044.1 | 4681 | UTR 3 | XP_016856533.1 |
Set Membership: |
HapMap |