Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCACAAAGACTCATGGGGCTT[C/T]CCTGGAAGAACTTCGTGCTTGATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612610 MIM: 612611 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE2B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LCE2B - late cornified envelope 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE2C - late cornified envelope 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178429.3 | 494 | UTR 3 | NP_848516.1 |