Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATATGACCCAAGTGTCCAGAA[C/T]TAATAAATGGCAGAGTGGATACAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601580 | ||||||||||||||||||||
Literature Links: |
CAPZA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPZA1 - capping actin protein of muscle Z-line alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006135.2 | Intron | NP_006126.1 | ||||
XM_011542225.2 | Intron | XP_011540527.1 | ||||
XM_017002424.1 | Intron | XP_016857913.1 |
ST7L - suppression of tumorigenicity 7 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |