Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGTTCTTTTTTACTGCTGAAT[A/G]GTTGTTCATGGTATGAGTGTATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600233 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GABRG3 - gamma-aminobutyric acid type A receptor gamma3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270873.1 | Intron | NP_001257802.1 | ||||
NM_033223.4 | Intron | NP_150092.2 | ||||
XM_011521430.2 | Intron | XP_011519732.1 | ||||
XM_011521431.2 | Intron | XP_011519733.1 | ||||
XM_017022058.1 | Intron | XP_016877547.1 | ||||
XM_017022059.1 | Intron | XP_016877548.1 | ||||
XM_017022060.1 | Intron | XP_016877549.1 |
Set Membership: |
HapMap |