Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTTAATAGAACTTTAAAGACA[G/T]CTGTTCTGACATTTATTTTAGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609787 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBAP1 - ubiquitin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171201.1 | Intron | NP_001164672.1 | ||||
NM_001171202.1 | Intron | NP_001164673.1 | ||||
NM_001171203.2 | Intron | NP_001164674.1 | ||||
NM_001171204.2 | Intron | NP_001164675.1 | ||||
NM_016525.4 | Intron | NP_057609.2 | ||||
XM_006716779.3 | Intron | XP_006716842.1 | ||||
XM_011517898.2 | Intron | XP_011516200.1 | ||||
XM_011517899.1 | Intron | XP_011516201.1 | ||||
XM_011517900.1 | Intron | XP_011516202.1 | ||||
XM_017014801.1 | Intron | XP_016870290.1 | ||||
XM_017014802.1 | Intron | XP_016870291.1 |