Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAACTACAAGTTCATTGCACATA[C/G]ACCACAGCTGACTGACTGAGTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602549 | ||||||||||||||||||||
Literature Links: |
PKN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PKN2 - protein kinase N2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320707.1 | Intron | NP_001307636.1 | ||||
NM_001320708.1 | Intron | NP_001307637.1 | ||||
NM_001320709.1 | Intron | NP_001307638.1 | ||||
NM_006256.3 | Intron | NP_006247.1 | ||||
XM_011541772.2 | Intron | XP_011540074.1 | ||||
XM_017001782.1 | Intron | XP_016857271.1 | ||||
XM_017001783.1 | Intron | XP_016857272.1 |