Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTTACCATTGCCATTTCCATAT[G/T]TGATCCTGTTACAACTTCATTTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927560 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927560 - uncharacterized LOC101927560 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927587 - uncharacterized LOC101927587 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL7 - tubulin tyrosine ligase like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024686.4 | 6107 | Intron | NP_078962.4 | |||
XM_005271208.2 | 6107 | UTR 3 | XP_005271265.1 | |||
XM_011542159.2 | 6107 | Intron | XP_011540461.1 | |||
XM_017002347.1 | 6107 | UTR 3 | XP_016857836.1 | |||
XM_017002348.1 | 6107 | Intron | XP_016857837.1 | |||
XM_017002349.1 | 6107 | UTR 3 | XP_016857838.1 |
Set Membership: |
HapMap |