Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGAAAAGGCACAGTAAACAGA[C/T]GGTGTCGTTATCTTATGTAGCCACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607144 MIM: 612552 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALG12 - ALG12, alpha-1,6-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024105.3 | Intron | NP_077010.1 | ||||
XM_011530371.2 | Intron | XP_011528673.1 | ||||
XM_017028936.1 | Intron | XP_016884425.1 | ||||
XM_017028937.1 | Intron | XP_016884426.1 |
ZBED4 - zinc finger BED-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014838.2 | Intron | NP_055653.2 |
Set Membership: |
HapMap |