Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103850 MIM: 615175 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDOA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ALDOA - aldolase, fructose-bisphosphate A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000034.3 | Intron | NP_000025.1 | ||||
NM_001127617.2 | Intron | NP_001121089.1 | ||||
NM_001243177.1 | Intron | NP_001230106.1 | ||||
NM_184041.2 | Intron | NP_908930.1 | ||||
NM_184043.2 | Intron | NP_908932.1 | ||||
XM_011545768.2 | Intron | XP_011544070.1 |
FAM57B - family with sequence similarity 57 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318504.1 | Intron | NP_001305433.1 | ||||
NM_031478.5 | Intron | NP_113666.2 | ||||
XM_005255613.3 | Intron | XP_005255670.1 | ||||
XM_005255614.3 | Intron | XP_005255671.1 | ||||
XM_005255615.2 | Intron | XP_005255672.1 | ||||
XM_017023749.1 | Intron | XP_016879238.1 | ||||
XM_017023750.1 | Intron | XP_016879239.1 | ||||
XM_017023751.1 | Intron | XP_016879240.1 | ||||
XM_017023752.1 | Intron | XP_016879241.1 | ||||
XM_017023753.1 | Intron | XP_016879242.1 | ||||
XM_017023754.1 | Intron | XP_016879243.1 |
Set Membership: |
HapMap |