Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609104 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO34 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXO34 - F-box protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017943.3 | Intron | NP_060413.2 | ||||
NM_152231.1 | Intron | NP_689417.1 | ||||
XM_005267791.4 | Intron | XP_005267848.1 | ||||
XM_006720185.3 | Intron | XP_006720248.1 | ||||
XM_017021391.1 | Intron | XP_016876880.1 | ||||
XM_017021392.1 | Intron | XP_016876881.1 | ||||
XM_017021393.1 | Intron | XP_016876882.1 |