Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATATTGTAAAAACTTTATTGAA[A/G]GCATCTGTCATATTAAGTGTTTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610577 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGAP12 - Rho GTPase activating protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270695.1 | Intron | NP_001257624.1 | ||||
NM_001270696.1 | Intron | NP_001257625.1 | ||||
NM_001270697.1 | Intron | NP_001257626.1 | ||||
NM_001270698.1 | Intron | NP_001257627.1 | ||||
NM_001270699.1 | Intron | NP_001257628.1 | ||||
NM_018287.6 | Intron | NP_060757.4 | ||||
XM_005252644.1 | Intron | XP_005252701.1 | ||||
XM_011519761.1 | Intron | XP_011518063.1 | ||||
XM_017016954.1 | Intron | XP_016872443.1 |
Set Membership: |
HapMap |