Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTAGCTTCAGATACCAATGCAT[A/G]TTACTCCTGACTTTGACATACTGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605730 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPAG6 - sperm associated antigen 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253854.1 | Intron | NP_001240783.1 | ||||
NM_001253855.1 | Intron | NP_001240784.1 | ||||
NM_012443.3 | Intron | NP_036575.1 | ||||
NM_172242.2 | Intron | NP_758442.1 | ||||
XM_005252645.1 | Intron | XP_005252702.1 | ||||
XM_005252646.1 | Intron | XP_005252703.1 |
Set Membership: |
HapMap |