Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTGAGTTGCAATGGTCTACACT[A/C]AAAGCCCCATGTTTCTGGCCCTCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608171 | |||||||||||||||||||||||
Literature Links: |
CACNA2D4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | Intron | NP_758952.4 | ||||
XM_011521041.2 | Intron | XP_011519343.1 |
LRTM2 - leucine rich repeats and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039029.2 | Intron | NP_001034118.1 | ||||
NM_001163925.1 | Intron | NP_001157397.1 | ||||
NM_001163926.1 | Intron | NP_001157398.1 | ||||
XM_011521015.1 | Intron | XP_011519317.1 | ||||
XM_017019848.1 | Intron | XP_016875337.1 |
Set Membership: |
HapMap |