Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGACCCCTTTTCTGAGTAAGGAAG[A/C]CCAGTTAAAATGTTAGCTTATTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605134 | ||||||||||||||||||||
Literature Links: |
PITPNC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PITPNC1 - phosphatidylinositol transfer protein, cytoplasmic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012417.3 | Intron | NP_036549.2 | ||||
NM_181671.2 | Intron | NP_858057.1 | ||||
XM_005257216.2 | Intron | XP_005257273.1 | ||||
XM_017024442.1 | Intron | XP_016879931.1 | ||||
XM_017024443.1 | Intron | XP_016879932.1 | ||||
XM_017024444.1 | Intron | XP_016879933.1 | ||||
XM_017024445.1 | Intron | XP_016879934.1 |