Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGACCTAGTATTTATTCTGGACC[A/G]AGATCTCATCGGCGAGTACCTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TDRD5 - tudor domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199085.1 | 863 | Silent Mutation | CCA,CCG | P,P 115 | NP_001186014.1 | |
NM_001199089.1 | 863 | Silent Mutation | CCA,CCG | P,P 115 | NP_001186018.1 | |
NM_001199091.1 | 863 | Silent Mutation | CCA,CCG | P,P 115 | NP_001186020.1 | |
NM_001199092.1 | 863 | Intron | NP_001186021.1 | |||
NM_173533.3 | 863 | Silent Mutation | CCA,CCG | P,P 115 | NP_775804.2 | |
XM_005244934.1 | 863 | Silent Mutation | CCA,CCG | P,P 115 | XP_005244991.1 | |
XM_005244935.4 | 863 | Silent Mutation | CCA,CCG | P,P 115 | XP_005244992.1 | |
XM_017000473.1 | 863 | Silent Mutation | CCA,CCG | P,P 115 | XP_016855962.1 |
Set Membership: |
HapMap |