Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601423 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLT8D2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLT8D2 - glycosyltransferase 8 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316967.1 | Intron | NP_001303896.1 | ||||
NM_031302.4 | Intron | NP_112592.1 | ||||
XM_011538793.2 | Intron | XP_011537095.1 | ||||
XM_017019999.1 | Intron | XP_016875488.1 |
TDG - thymine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003211.4 | Intron | NP_003202.3 | ||||
XM_005269125.1 | Intron | XP_005269182.1 | ||||
XM_011538714.1 | Intron | XP_011537016.1 | ||||
XM_011538715.1 | Intron | XP_011537017.1 |
Set Membership: |
HapMap |