Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGAAGGAAGGAAATTAGCAT[C/T]ACACCTTTCTTTTATGAGCCTGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613802 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLEC PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLEC - malectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303627.1 | Intron | NP_001290556.1 | ||||
NM_001303628.1 | Intron | NP_001290557.1 | ||||
NM_014730.3 | Intron | NP_055545.1 | ||||
XM_011539031.1 | Intron | XP_011537333.1 | ||||
XM_011539032.1 | Intron | XP_011537334.1 |