Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGAGGGGCACACATCCTTCCT[A/G]GAAGAAGCTGGGTAGGCTCTATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602059 MIM: 610745 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISLR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ISLR - immunoglobulin superfamily containing leucine rich repeat | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRA6 - stimulated by retinoic acid 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142617.1 | Intron | NP_001136089.1 | ||||
NM_001142618.1 | Intron | NP_001136090.1 | ||||
NM_001142619.1 | Intron | NP_001136091.1 | ||||
NM_001142620.1 | Intron | NP_001136092.1 | ||||
NM_001199040.1 | Intron | NP_001185969.1 | ||||
NM_001199041.1 | Intron | NP_001185970.1 | ||||
NM_001199042.1 | Intron | NP_001185971.1 | ||||
NM_022369.3 | Intron | NP_071764.3 | ||||
XM_011521883.1 | Intron | XP_011520185.1 | ||||
XM_011521884.1 | Intron | XP_011520186.1 | ||||
XM_011521885.2 | Intron | XP_011520187.1 | ||||
XM_017022478.1 | Intron | XP_016877967.1 | ||||
XM_017022479.1 | Intron | XP_016877968.1 | ||||
XM_017022480.1 | Intron | XP_016877969.1 |