Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCAGTGAGATATTGAAAGATTC[C/T]AAGTCTAGCTGACGAGAGCCTGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606029 MIM: 607153 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPSF3 - cleavage and polyadenylation specific factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321833.1 | Intron | NP_001308762.1 | ||||
NM_001321834.1 | Intron | NP_001308763.1 | ||||
NM_001321835.1 | Intron | NP_001308764.1 | ||||
NM_001321836.1 | Intron | NP_001308765.1 | ||||
NM_016207.3 | Intron | NP_057291.1 |
ITGB1BP1 - integrin subunit beta 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |