Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCTGCTCCGCGTTGTTTAGAT[A/T]GAGATGCTGCCTCACGGTCCTGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142620 MIM: 610434 | ||||||||||||||||||||
Literature Links: |
ABHD16A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD16A - abhydrolase domain containing 16A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177515.1 | 1428 | Missense Mutation | AAT,TAT | N,Y 375 | NP_001170986.1 | |
NM_021160.2 | 1428 | Missense Mutation | AAT,TAT | N,Y 408 | NP_066983.1 |
LY6G5C - lymphocyte antigen 6 complex, locus G5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |