Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGCATCAAATATTATGTAGTCA[G/T]AGCAGGCTTTTTAAAGCTAGTAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605649 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP44 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP44 - centrosomal protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040157.2 | Intron | NP_001035247.1 | ||||
NM_001145314.1 | Intron | NP_001138786.1 | ||||
XM_011532285.2 | Intron | XP_011530587.1 | ||||
XM_011532286.2 | Intron | XP_011530588.1 | ||||
XM_011532287.2 | Intron | XP_011530589.1 | ||||
XM_017008654.1 | Intron | XP_016864143.1 | ||||
XM_017008655.1 | Intron | XP_016864144.1 | ||||
XM_017008656.1 | Intron | XP_016864145.1 | ||||
XM_017008657.1 | Intron | XP_016864146.1 | ||||
XM_017008658.1 | Intron | XP_016864147.1 | ||||
XM_017008659.1 | Intron | XP_016864148.1 | ||||
XM_017008660.1 | Intron | XP_016864149.1 |
FBXO8 - F-box protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |