Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616213 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF292 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF292 - zinc finger protein 292 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015021.1 | Intron | NP_055836.1 | ||||
XM_005248697.4 | Intron | XP_005248754.1 | ||||
XM_011535625.2 | Intron | XP_011533927.1 | ||||
XM_017010575.1 | Intron | XP_016866064.1 | ||||
XM_017010576.1 | Intron | XP_016866065.1 | ||||
XM_017010577.1 | Intron | XP_016866066.1 | ||||
XM_017010578.1 | Intron | XP_016866067.1 | ||||
XM_017010579.1 | Intron | XP_016866068.1 | ||||
XM_017010580.1 | Intron | XP_016866069.1 | ||||
XM_017010581.1 | Intron | XP_016866070.1 | ||||
XM_017010582.1 | Intron | XP_016866071.1 | ||||
XM_017010583.1 | Intron | XP_016866072.1 | ||||
XM_017010584.1 | Intron | XP_016866073.1 |
Set Membership: |
HapMap |