Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGATGATTCTTGGTGTGGGCTG[A/T]GGGAGCTCCTGCGGTGTAGTTTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603076 | ||||||||||||||||||||
Literature Links: |
ABCG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCG1 - ATP binding cassette subfamily G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004915.3 | Intron | NP_004906.3 | ||||
NM_016818.2 | Intron | NP_058198.2 | ||||
NM_207174.1 | Intron | NP_997057.1 | ||||
NM_207627.1 | Intron | NP_997510.1 | ||||
NM_207628.1 | Intron | NP_997511.1 | ||||
NM_207629.1 | Intron | NP_997512.1 | ||||
XM_011529806.1 | Intron | XP_011528108.1 | ||||
XM_011529807.2 | Intron | XP_011528109.1 |