Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTTTGCTTCCTATCCCTGAGAA[C/G]TTATATTCCACAGGACTAGAGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610638 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGSF5 - immunoglobulin superfamily member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080444.1 | 676 | Intron | NP_001073913.1 | |||
XM_011529472.1 | 676 | UTR 5 | XP_011527774.1 | |||
XM_011529473.1 | 676 | UTR 5 | XP_011527775.1 | |||
XM_017028279.1 | 676 | UTR 5 | XP_016883768.1 | |||
XM_017028280.1 | 676 | UTR 5 | XP_016883769.1 |
Set Membership: |
HapMap |