Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTCAGGAGGCATCAAAGGAGC[A/G]TTTGTTACAGGCTTTAGTAGATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194500 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF2 - zinc finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017396.2 | Intron | NP_001017396.1 | ||||
NM_001282398.1 | Intron | NP_001269327.1 | ||||
NM_001291604.1 | Intron | NP_001278533.1 | ||||
NM_001291605.1 | Intron | NP_001278534.1 | ||||
NM_021088.3 | Intron | NP_066574.2 |
Set Membership: |
HapMap |