Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGGAGGTGAAGCAAACTGAAGA[G/T]TTAAGTATGAGTTCTAGCTTTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603525 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NMI - N-myc and STAT interactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004688.2 | Intron | NP_004679.2 | ||||
XM_005246941.2 | Intron | XP_005246998.1 | ||||
XM_017005247.1 | Intron | XP_016860736.1 |
RBM43 - RNA binding motif protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |