Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGGGACCCACCCCAAGGTGCT[C/T]TTAGCTCAAGGTCATGCCTGGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611624 | ||||||||||||||||||||
Literature Links: |
NECAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NECAP2 - NECAP endocytosis associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145277.1 | Intron | NP_001138749.1 | ||||
NM_001145278.1 | Intron | NP_001138750.1 | ||||
NM_018090.4 | Intron | NP_060560.1 | ||||
XM_011541767.2 | Intron | XP_011540069.1 | ||||
XM_017001721.1 | Intron | XP_016857210.1 |
SPATA21 - spermatogenesis associated 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |