Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAAATTTGACTTGTGATTCTTT[A/G]TACATGTTTCATTGTTTGTGTTGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159553 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MNDA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MNDA - myeloid cell nuclear differentiation antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002432.1 | Intron | NP_002423.1 |
OR6N1 - olfactory receptor family 6 subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005185.1 | Intron | NP_001005185.1 | ||||
XM_017000325.1 | Intron | XP_016855814.1 | ||||
XM_017000326.1 | Intron | XP_016855815.1 | ||||
XM_017000327.1 | Intron | XP_016855816.1 |
Set Membership: |
HapMap |