Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCCACCCATGACTGCAGACATG[A/T]CCCTATAAGTCATGGCCCCAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1C3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SULT1C3 - sulfotransferase family 1C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008743.2 | Intron | NP_001008743.1 | ||||
NM_001320878.1 | Intron | NP_001307807.1 | ||||
XM_017004153.1 | Intron | XP_016859642.1 | ||||
XM_017004154.1 | Intron | XP_016859643.1 | ||||
XM_017004155.1 | Intron | XP_016859644.1 |
Set Membership: |
HapMap |