Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTTGGGGAAATTGAGGCACA[G/T]AAATGTGGTAACTTGCCCAAGGCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612373 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC29A3 - solute carrier family 29 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174098.1 | Intron | NP_001167569.1 | ||||
NM_018344.5 | Intron | NP_060814.4 | ||||
XM_006717910.3 | Intron | XP_006717973.1 | ||||
XM_017016377.1 | Intron | XP_016871866.1 | ||||
XM_017016378.1 | Intron | XP_016871867.1 |
Set Membership: |
HapMap |