Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTACTGTTAGAGGTAGAGAAAT[A/C]CAATATCATAAAGAAAGATGTGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601799 | ||||||||||||||||||||
Literature Links: |
LOC101927730 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927730 - uncharacterized LOC101927730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINB9 - serpin family B member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004155.5 | 2385 | UTR 3 | NP_004146.1 | |||
XM_005249184.4 | 2385 | UTR 3 | XP_005249241.1 | |||
XM_011514678.2 | 2385 | UTR 3 | XP_011512980.1 | |||
XM_017010942.1 | 2385 | UTR 3 | XP_016866431.1 | |||
XM_017010943.1 | 2385 | UTR 3 | XP_016866432.1 |