Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAAAAGAATAAAAAACAGAAAA[C/T]TTCCTACAAGAGAGGAGTCAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614234 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIFO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIFO - primary cilia formation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300831.1 | Intron | NP_001287760.1 | ||||
NM_181643.5 | Intron | NP_857594.2 | ||||
XM_005270472.1 | Intron | XP_005270529.1 | ||||
XM_017000322.1 | Intron | XP_016855811.1 |