Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTTCTTATATGAGCTTGTTAT[G/T]ATAGTAAGTTATCTCAACAAATGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107930 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDC PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDC - dopa decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000790.3 | Intron | NP_000781.1 | ||||
NM_001082971.1 | Intron | NP_001076440.1 | ||||
NM_001242886.1 | Intron | NP_001229815.1 | ||||
NM_001242887.1 | Intron | NP_001229816.1 | ||||
NM_001242888.1 | Intron | NP_001229817.1 | ||||
NM_001242889.1 | Intron | NP_001229818.1 | ||||
NM_001242890.1 | Intron | NP_001229819.1 | ||||
XM_005271745.4 | Intron | XP_005271802.1 | ||||
XM_011515161.2 | Intron | XP_011513463.2 |
Set Membership: |
HapMap |