Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATATGACTCAAGAAATATCCCA[G/T]AACTAAGCAAAGAGGTCACTGAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604650 | ||||||||||||||||||||
Literature Links: |
IFIT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFIT3 - interferon induced protein with tetratricopeptide repeats 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031683.3 | Intron | NP_001026853.1 | ||||
NM_001289758.1 | Intron | NP_001276687.1 | ||||
NM_001289759.1 | Intron | NP_001276688.1 | ||||
NM_001549.5 | Intron | NP_001540.2 |