Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTTGGAAATCATCTCAGCTTC[C/T]TAAAACACAAAACAAAAGTTTAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604873 MIM: 611707 | ||||||||||||||||||||
Literature Links: |
MPZL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPZL2 - myelin protein zero like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005797.3 | Intron | NP_005788.1 | ||||
NM_144765.2 | Intron | NP_658911.1 |
MPZL3 - myelin protein zero like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |