Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACATGTTCAAACCAGAAGTATT[C/T]GCTGCCCCCAGCTCTAAGAATCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616587 MIM: 182180 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM118B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM118B - family with sequence similarity 118 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024556.3 | 3619 | Intron | NP_078832.1 | |||
XM_005271665.3 | 3619 | Intron | XP_005271722.1 | |||
XM_011542977.2 | 3619 | Intron | XP_011541279.1 | |||
XM_011542978.2 | 3619 | Intron | XP_011541280.1 | |||
XM_017018285.1 | 3619 | Intron | XP_016873774.1 | |||
XM_017018286.1 | 3619 | Intron | XP_016873775.1 |
SRPRA - SRP receptor alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177842.1 | 3619 | Intron | NP_001171313.1 | |||
NM_003139.3 | 3619 | Intron | NP_003130.2 | |||
XM_017018179.1 | 3619 | UTR 3 | XP_016873668.1 |
Set Membership: |
HapMap |